Gene Transcripts

Gene Symbol
CNPPD1
Entrez Gene ID
27013
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
C2orf24, CGI-57
Chromosome Location
Chr. 2: 219171897 - 219178174 on Build GRCh38
UniGene ID
Hs.4973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321389.1 NP_001308318.1 6, 7 944
NM_001321390.1 NP_001308319.1 6, 7 589
NM_001321391.1 NP_001308320.1 6, 7 623
NM_015680.5 NP_056495.3 5, 6 681
GenBank mRNA AF151815.1 - 6, 7 848
AK075274.1 - 5, 6 637
AK314975.1 - 5, 6 624
AY007160.1 - 5, 6 654
BC001393.2 - 5, 6 604 3051397
BC012821.2 - 5, 6 653 2958621
BI912613.1 - 6, 7 590
CR457401.1 - 5, 6 516

View Full Product Details