Gene Transcripts

Gene Symbol
SLC39A1
Entrez Gene ID
27173
Gene Name
solute carrier family 39 member 1
Gene Aliases
ZIP1, ZIRTL
Chromosome Location
Chr. 1: 153959099 - 153968184 on Build GRCh38
UniGene ID
Hs.743291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271957.1 NP_001258886.1 4 1252
NM_001271958.1 NP_001258887.1 4 1089
NM_001271959.1 NP_001258888.1 4 1258
NM_001271960.1 NP_001258889.1 5 1150
NM_001271961.1 NP_001258890.1 3 958
NM_014437.4 NP_055252.2 5 1396
GenBank mRNA AF132942.1 - 3 946
AF151829.1 - 4 967
AJ243649.1 - 4 983
AJ271671.1 - 4 997
AK074943.1 - 4 1206
AK293389.1 - 3 866
AK297983.1 - 5 964
AK300244.1 - 5 956
BC002563.2 - 4 971 3138575
BC003152.2 - 3 943 2958651
BC007886.2 - 4 968 4122086
BC014303.2 - 5 1376 3942586

View Full Product Details