Gene Transcripts

Gene Symbol
NFU1
Entrez Gene ID
27247
Gene Name
NFU1 iron-sulfur cluster scaffold
Gene Aliases
CGI-33, HIRIP, HIRIP5, MMDS1, NIFUC, Nfu, NifU
Chromosome Location
Chr. 2: 69396113 - 69438123 on Build GRCh38
UniGene ID
Hs.430439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002755.2 NP_001002755.1 7 825
NM_001002756.2 NP_001002756.1 6 779
NM_015700.3 NP_056515.2 7 915
NR_045631.1 6 689
XM_017003808.1 XP_016859297.1 7 628
GenBank mRNA AA625581.1 - 2 270
AF132967.1 - 7 627
AI129426.1 - 2 279
AJ132584.1 - 7 703
AK300700.1 - 7 634
AK314004.1 - 7 658
AY286306.1 - 7 672
AY286307.1 - 7 762
AY335194.1 - 7 818
BC113692.1 - 7 787 8322744
BC113694.1 - 7 787 8322746
BU178674.1 - 6 521
BU507991.1 - 7 630
BX538347.1 - 7 690

View Full Product Details