Gene Transcripts

Gene Symbol
LSM1
Entrez Gene ID
27257
Gene Name
LSM1 homolog, mRNA degradation associated
Gene Aliases
CASM, YJL124C
Chromosome Location
Chr. 8: 38163321 - 38176730 on Build GRCh38
UniGene ID
Hs.425311
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014462.2 NP_055277.1 3 556
NR_045493.1 3 278
GenBank mRNA AF000177.1 - 3 310
AF147348.1 - 3 199
AJ238094.1 - 3 334
AK312159.1 - 3 309
BC001767.1 - 3 243 3353833
BP376421.1 - 3 519
BU941962.1 - 3 281
KU178606.1 - 3 146

View Full Product Details