Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014012.5 | NP_054731.2 | 4 | 768 | |
XM_005260404.1 | XP_005260461.1 | 4 | 792 | ||
XM_011528795.1 | XP_011527097.1 | 4 | 786 | ||
XM_017027833.1 | XP_016883322.1 | 4 | 626 | ||
GenBank mRNA | AF084465.1 | - | 3 | 542 | |
AF152863.1 | - | 4 | 750 | ||
AK313060.1 | - | 4 | 768 | ||
BC039813.1 | - | 4 | 754 | 5220518 | |
BT020078.1 | - | 3 | 470 | ||
BT020079.1 | - | 3 | 470 |