Gene Transcripts
- Gene Symbol
- WDR91
- Entrez Gene ID
- 29062
- Gene Name
- WD repeat domain 91
- Gene Aliases
- HSPC049, SORF-1, SORF1
- Chromosome Location
- Chr. 7: 135183838 - 135211564 on Build GRCh38
- UniGene ID
- Hs.459858
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014149.3 |
NP_054868.3
|
5, 6 |
793 |
|
XM_011516083.2 |
XP_011514385.1
|
5, 6 |
763 |
|
XM_011516086.2 |
XP_011514388.1
|
5, 6 |
763 |
|
XM_017012070.1 |
XP_016867559.1
|
5, 6 |
763 |
|
GenBank mRNA |
AF161534.1 |
- |
5, 6 |
735 |
|
AK001606.1 |
- |
5, 6 |
748 |
|
AK022935.1 |
- |
5, 6 |
737 |
|
AL136940.1 |
- |
5, 6 |
644 |
|
BC009939.2 |
- |
3, 4 |
251 |
|
BC017246.2 |
- |
5, 6 |
732 |
4890904 |
BC063394.1 |
- |
5, 6 |
729 |
|
BX640957.1 |
- |
5, 6 |
764 |
|
CR533561.1 |
- |
4, 5 |
575 |
|
View Full Product Details