Gene Transcripts

Gene Symbol
ST8SIA5
Entrez Gene ID
29906
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5
Gene Aliases
SIAT8-E, SIAT8E, ST8SiaV
Chromosome Location
Chr. 18: 46679118 - 46757169 on Build GRCh38
UniGene ID
Hs.465025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001307986.1 NP_001294915.1 3 958
NM_013305.5 NP_037437.2 2 850
XM_017025715.1 XP_016881204.1 4 346
XM_017025716.1 XP_016881205.1 2 491
GenBank mRNA AK056270.1 - 2 850
AK130126.1 - 4 405
AK293608.1 - 3 598
AK310764.1 - 3 599
BC108910.1 - 2 400 40011279
BC108911.1 - 2 400 40011281
CR457037.1 - 2 189
U91641.1 - 2 236

View Full Product Details