Gene Transcripts
- Gene Symbol
- C18orf8
- Entrez Gene ID
- 29919
- Gene Name
- chromosome 18 open reading frame 8
- Gene Aliases
- HsT2591, MIC1, Mic-1
- Chromosome Location
- Chr. 18: 23503470 - 23531807 on Build GRCh38
- UniGene ID
- Hs.464779
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001276342.1 |
NP_001263271.1
|
2, 3 |
338 |
|
NM_001318709.1 |
NP_001305638.1
|
2, 3 |
338 |
|
NM_013326.4 |
NP_037458.3
|
2, 3 |
338 |
|
GenBank mRNA |
AF143536.1 |
- |
1, 2 |
85 |
|
AK057192.1 |
- |
2, 3 |
299 |
|
AK296230.1 |
- |
2, 3 |
289 |
|
BC002950.1 |
- |
2, 3 |
263 |
3538017 |
BC008305.1 |
- |
2, 3 |
270 |
3537084 |
CN481504.1 |
- |
2, 3 |
310 |
|
DC383182.1 |
- |
2, 3 |
322 |
|
View Full Product Details