Gene Transcripts
- Gene Symbol
- SAR1B
- Entrez Gene ID
- 51128
- Gene Name
- secretion associated Ras related GTPase 1B
- Gene Aliases
- ANDD, CMRD, GTBPB, SARA2
- Chromosome Location
- Chr. 5: 134601149 - 134632843 on Build GRCh38
- UniGene ID
- Hs.432984
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033503.2 |
NP_001028675.1
|
4, 5 |
425 |
|
NM_016103.3 |
NP_057187.1
|
3, 4 |
306 |
|
XM_017009521.1 |
XP_016865010.1
|
3, 4 |
313 |
|
GenBank mRNA |
AF087850.1 |
- |
3, 4 |
343 |
|
AF092130.1 |
- |
3, 4 |
322 |
|
AK223452.1 |
- |
3, 4 |
291 |
|
AL710660.1 |
- |
4, 5 |
407 |
|
BC002847.2 |
- |
4, 5 |
356 |
3636540 |
BC093034.1 |
- |
3, 4 |
306 |
30528518 |
CD048852.1 |
- |
4, 5 |
433 |
|
View Full Product Details