Gene Transcripts

Gene Symbol
UFM1
Entrez Gene ID
51569
Gene Name
ubiquitin fold modifier 1
Gene Aliases
BM-002, C13orf20
Chromosome Location
Chr. 13: 38349771 - 38363007 on Build GRCh38
UniGene ID
Hs.643655
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286703.1 NP_001273632.1 4 279
NM_001286704.1 NP_001273633.1 4 457
NM_001286705.1 NP_001273634.1 4 285
NM_001286706.1 NP_001273635.1 4 285
NM_016617.3 NP_057701.1 4 285
NR_104584.1 5 346
GenBank mRNA AB154404.1 - 3 136
AF208844.1 - 4 175
AK095490.1 - 4 205
AK307111.1 - 4 205
AK309914.1 - 4 171
AL515797.3 - 3 137
BC005193.1 - 4 202 3829206
BG533062.1 - 4 378
BG540094.1 - 4 173
BI458294.1 - 4 256
BI761948.1 - 4 162
BP277186.1 - 4 285
BU589105.1 - 5 262
CR457189.1 - 3 136
DA483023.1 - 4 205

View Full Product Details