Gene Transcripts
- Gene Symbol
- SRRT
- Entrez Gene ID
- 51593
- Gene Name
- serrate, RNA effector molecule
- Gene Aliases
- ARS2, ASR2, serrate
- Chromosome Location
- Chr. 7: 100874971 - 100888664 on Build GRCh38
- UniGene ID
- Hs.111801
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128852.1 |
NP_001122324.1
|
14, 15 |
2077 |
|
NM_001128853.1 |
NP_001122325.1
|
14, 15 |
2080 |
|
NM_001128854.1 |
NP_001122326.1
|
14, 15 |
2077 |
|
NM_015908.5 |
NP_056992.4
|
14, 15 |
2080 |
|
XM_005250405.2 |
XP_005250462.1
|
14, 15 |
1937 |
|
XM_005250406.2 |
XP_005250463.1
|
14, 15 |
1934 |
|
XM_005250407.2 |
XP_005250464.1
|
14, 15 |
1937 |
|
XM_005250408.2 |
XP_005250465.1
|
14, 15 |
1934 |
|
XM_017012290.1 |
XP_016867779.1
|
15, 16 |
2262 |
|
XM_017012291.1 |
XP_016867780.1
|
15, 16 |
2259 |
|
GenBank mRNA |
AF082871.1 |
- |
4, 5 |
488 |
|
AF112994.1 |
- |
1, 2 |
263 |
|
AF248955.1 |
- |
14, 15 |
2043 |
|
AK296131.1 |
- |
8, 9 |
1046 |
|
AK302820.1 |
- |
14, 15 |
1969 |
|
AL096723.1 |
- |
14, 15 |
2031 |
|
BC000082.2 |
- |
5, 6 |
601 |
|
BC069249.1 |
- |
12, 13 |
1621 |
|
BC109117.1 |
- |
14, 15 |
1971 |
40035609 |
BQ059529.1 |
- |
2, 3 |
376 |
|
View Full Product Details