Gene Transcripts

Gene Symbol
NUDT9
Entrez Gene ID
53343
Gene Name
nudix hydrolase 9
Gene Aliases
NUDT10
Chromosome Location
Chr. 4: 87420002 - 87459454 on Build GRCh38
UniGene ID
Hs.149500
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001248011.1 NP_001234940.1 2 625
NM_024047.4 NP_076952.1 2 625
NM_198038.2 NP_932155.1 2 267
XM_011532031.2 XP_011530333.1 2 628
XM_011532032.2 XP_011530334.1 2 2804
GenBank mRNA AF265341.1 - 2 266
AF273028.1 - 2 589
AK027295.1 - 2 615
AK074845.1 - 2 267
AK075408.1 - 2 545
AK308028.1 - 2 392
AY026252.1 - 2 620
AY359123.1 - 2 319
BC000542.2 - 2 385 3163944
BI552154.1 - 265
BM544951.1 - 2 637

View Full Product Details