Gene Transcripts

Gene Symbol
MIS18A
Entrez Gene ID
54069
Gene Name
MIS18 kinetochore protein A
Gene Aliases
B28, C21orf45, C21orf46, FASP1, MIS18alpha, hMis18alpha
Chromosome Location
Chr. 21: 32244759 - 32283954 on Build GRCh38
UniGene ID
Hs.190518
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018944.2 NP_061817.1 2 418
XM_017028400.1 XP_016883889.1 2 509
XM_017028401.1 XP_016883890.1 2 509
XM_017028402.1 XP_016883891.1 2 117
GenBank mRNA AF231921.1 - 2 390
AF387845.1 - 2 388
AK075281.1 - 2 390
AK312073.1 - 2 509
BC042917.1 - 2 394 5934336
BU616573.1 - 2 421
CB959520.1 - 467

View Full Product Details