Gene Transcripts

Gene Symbol
ZNHIT6
Entrez Gene ID
54680
Gene Name
zinc finger HIT-type containing 6
Gene Aliases
BCD1, C1orf181, NY-BR-75
Chromosome Location
Chr. 1: 85649423 - 85708447 on Build GRCh38
UniGene ID
Hs.5111
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170670.1 NP_001164141.1 4 797
NM_017953.3 NP_060423.3 3 914
XM_011541614.2 XP_011539916.1 3 928
GenBank mRNA AF308296.1 - 3 561
AK000736.1 - 3 286
AK000767.1 - 3 823
AK298149.1 - 4 797
AK314566.1 - 3 842
AL442074.1 - 3 899
BC026236.1 - 3 901 4664103
BC110898.1 - 3 832 6012059

View Full Product Details