Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040125.1 | NP_001035214.1 | 4 | 1059 | |
NM_001040126.1 | NP_001035215.1 | 4 | 447 | ||
NM_001287531.1 | NP_001274460.1 | 4 | 416 | ||
NM_017765.2 | NP_060235.2 | 5 | 689 | ||
NR_109848.1 | 5 | 689 | |||
XM_005245916.2 | XP_005245973.1 | 4 | 472 | ||
XM_006710710.2 | XP_006710773.1 | 5 | 582 | ||
XM_006710711.2 | XP_006710774.1 | 4 | 441 | ||
XM_011541634.2 | XP_011539936.1 | 5 | 641 | ||
GenBank mRNA | AK000327.1 | - | 5 | 610 | |
AK125580.1 | - | 4 | 977 | ||
AK130171.1 | - | 5 | 607 | ||
AK130175.1 | - | 5 | 609 | ||
AK172745.1 | - | 4 | 334 | ||
BC015324.1 | - | 4 | 352 | 4426038 | |
KU178659.1 | - | 2 | 142 | ||
KU178660.1 | - | 2 | 111 |