Gene Transcripts

Gene Symbol
DUS2
Entrez Gene ID
54920
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, SMM1, URLC8
Chromosome Location
Chr. 16: 68022944 - 68079323 on Build GRCh38
UniGene ID
Hs.534460
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271762.1 NP_001258691.1 15 1618
NM_001271763.1 NP_001258692.1 14 1593
NM_017803.4 NP_060273.1 16 1698
GenBank mRNA AB101210.1 - 16 1362
AK000406.1 - 15 1265
AK290578.1 - 16 1400
AK296663.1 - 14 1245
BC006527.2 - 16 1341 2988128

View Full Product Details