Gene Transcripts

Gene Symbol
SAGE1
Entrez Gene ID
55511
Gene Name
sarcoma antigen 1
Gene Aliases
CT14, SAGE
Chromosome Location
Chr. X: 135893607 - 135913061 on Build GRCh38
UniGene ID
Hs.195292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018666.2 NP_061136.2 10 1217
XM_017029621.1 XP_016885110.1 10 1225
XM_017029622.1 XP_016885111.1 10 1136
XM_017029623.1 XP_016885112.1 10 1132
XM_017029624.1 XP_016885113.1 9 1039
XM_017029625.1 XP_016885114.1 10 1132
XM_017029626.1 XP_016885115.1 4 341
GenBank mRNA AJ278111.1 - 10 1217
AK310412.1 - 9 1173
CD358947.1 - 2 55

View Full Product Details