Gene Transcripts
- Gene Symbol
- NPLOC4
- Entrez Gene ID
- 55666
- Gene Name
- NPL4 homolog, ubiquitin recognition factor
- Gene Aliases
- NPL4
- Chromosome Location
- Chr. 17: 81556885 - 81637153 on Build GRCh38
- UniGene ID
- Hs.464333
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017921.3 |
NP_060391.2
|
3, 4 |
443 |
|
XM_011524979.1 |
XP_011523281.1
|
3, 4 |
438 |
|
XM_011524980.1 |
XP_011523282.1
|
3, 4 |
438 |
|
XM_011524981.1 |
XP_011523283.1
|
3, 4 |
438 |
|
XM_011524982.2 |
XP_011523284.1
|
3, 4 |
438 |
|
GenBank mRNA |
AB040932.1 |
- |
3, 4 |
350 |
|
AK000664.1 |
- |
3, 4 |
352 |
|
AK023046.1 |
- |
1, 2 |
79 |
|
AK023272.1 |
- |
1 |
22 |
|
AK294473.1 |
- |
3, 4 |
369 |
|
BC025930.1 |
- |
3, 4 |
384 |
5017718 |
DC322260.1 |
- |
3, 4 |
402 |
|
HY116973.1 |
- |
3, 4 |
445 |
|
View Full Product Details