Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242617.1 | NP_001229546.1 | 3 | 347 | |
NM_001242618.1 | NP_001229547.1 | 3 | 308 | ||
NM_018698.4 | NP_061168.2 | 4 | 437 | ||
XM_011530989.2 | XP_011529291.1 | 3 | 350 | ||
XM_017029648.1 | XP_016885137.1 | 3 | 285 | ||
GenBank mRNA | AF201942.1 | - | 3 | 314 | |
AF212223.1 | - | 4 | 299 | ||
AF246127.1 | - | 2 | 170 | ||
AJ277591.1 | - | 2 | 170 | ||
AJ278323.1 | - | 3 | 308 | ||
AK023289.1 | - | 4 | 437 | ||
BC014888.1 | - | 3 | 301 | ||
BC120984.1 | - | 4 | 426 | 40116082 | |
BC120985.1 | - | 4 | 426 | 40116088 | |
DA442845.1 | - | 3 | 349 |