Gene Transcripts

Gene Symbol
CTTNBP2NL
Entrez Gene ID
55917
Gene Name
CTTNBP2 N-terminal like
Gene Aliases
Chromosome Location
Chr. 1: 112391085 - 112461164 on Build GRCh38
UniGene ID
Hs.744100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018704.2 NP_061174.1 6 1254
XM_011541781.2 XP_011540083.1 6 1325
XM_017001806.1 XP_016857295.1 6 1692
GenBank mRNA AK022544.1 - 5 1175
AK023340.1 - 1348
BC016029.1 - 6 1194 3921313

View Full Product Details