Gene Transcripts

Gene Symbol
PCDHGB6
Entrez Gene ID
56100
Gene Name
protocadherin gamma subfamily B, 6
Gene Aliases
PCDH-GAMMA-B6
Chromosome Location
Chr. 5: 141408203 - 141512979 on Build GRCh38
UniGene ID
Hs.368160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018926.2 NP_061749.1 1 1139
NM_032100.1 NP_115271.1 1 1139
GenBank mRNA AF152335.1 - 1 1139
AF152522.1 - 1 1139

View Full Product Details