Gene Transcripts

Gene Symbol
TMEM9B
Entrez Gene ID
56674
Gene Name
TMEM9 domain family member B
Gene Aliases
C11orf15
Chromosome Location
Chr. 11: 8947201 - 8965011 on Build GRCh38
UniGene ID
Hs.501853
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286094.1 NP_001273023.1 3 715
NM_001286095.1 NP_001273024.1 4 364
NM_020644.2 NP_065695.1 4 807
GenBank mRNA AB097017.1 - 4 432
AK074677.1 - 4 425
AK300712.1 - 4 287
AK310723.1 - 3 346
AK313065.1 - 4 437
AK316216.1 - 3 346
AY359069.1 - 4 450
BC015884.1 - 3 342 3909465
BC040124.1 - 4 489 6053804
DC314550.1 - 4 364

View Full Product Details