Gene Transcripts

Gene Symbol
C8orf4
Entrez Gene ID
56892
Gene Name
chromosome 8 open reading frame 4
Gene Aliases
TC-1, TC1
Chromosome Location
Chr. 8: 40153468 - 40155308 on Build GRCh38
UniGene ID
Hs.591849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020130.4 NP_064515.1 1 149
GenBank mRNA AF268037.1 - 1 133
AK026240.1 - 1 141
BC020623.1 - 1 135 4718061
BC021672.1 - 1 153 3682270

View Full Product Details