Gene Transcripts

Gene Symbol
TRPC7
Entrez Gene ID
57113
Gene Name
transient receptor potential cation channel subfamily C member 7
Gene Aliases
TRP7
Chromosome Location
Chr. 5: 136213311 - 136365476 on Build GRCh38
UniGene ID
Hs.591263
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167576.1 NP_001161048.1 6 1829
NM_001167577.1 NP_001161049.1 7 1994
NM_020389.2 NP_065122.1 8 2177
GenBank mRNA AJ272034.1 - 7 1955
AJ421783.1 - 7 1955
AJ549088.1 - 6 1772
AJ549089.1 - 6 1790
AJ549090.1 - 4 1390
AY167926.1 - 6 1787
AY167927.1 - 5 1607
BC128185.1 - 7 1955 40113370
BC128186.1 - 7 1971

View Full Product Details