Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020675.3 | NP_065726.1 | 4, 5 | 477 | |
XM_011511516.2 | XP_011509818.1 | 4, 5 | 486 | ||
GenBank mRNA | AF225416.1 | - | 4, 5 | 459 | |
AK291093.1 | - | 4, 5 | 488 | ||
AY456388.1 | - | 3, 4 | 346 | ||
BC022255.1 | - | 4, 5 | 477 | 4738187 | |
CR457197.1 | - | 3, 4 | 346 |