Gene Transcripts

Gene Symbol
COG6
Entrez Gene ID
57511
Gene Name
component of oligomeric golgi complex 6
Gene Aliases
CDG2L, COD2, SHNS
Chromosome Location
Chr. 13: 39655627 - 39791665 on Build GRCh38
UniGene ID
Hs.507805
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145079.1 NP_001138551.1 13 1287
NM_020751.2 NP_065802.1 13 1287
NR_026745.1 14 1352
XM_011535168.1 XP_011533470.1 13 1237
GenBank mRNA AB032960.1 - 13 1177
AF116827.1 - 14 1235
AK026638.1 - 12 954
AK294443.1 - 13 1287
BC051723.1 - 13 1237 5263056
CR627406.1 - 13 1230

View Full Product Details