Gene Transcripts
- Gene Symbol
- HACE1
- Entrez Gene ID
- 57531
- Gene Name
- HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1
- Gene Aliases
- SPPRS
- Chromosome Location
- Chr. 6: 104682670 - 104859919 on Build GRCh38
- UniGene ID
- Hs.434340
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321080.1 |
NP_001308009.1
|
2, 3 |
418 |
|
NM_001321083.1 |
NP_001308012.1
|
2, 3 |
233 |
|
NM_001321084.1 |
NP_001308013.1
|
2, 3 |
233 |
|
NM_020771.3 |
NP_065822.2
|
2, 3 |
418 |
|
NR_104424.1 |
|
2, 3 |
418 |
|
XM_017011113.1 |
XP_016866602.1
|
3, 4 |
417 |
|
XM_017011114.1 |
XP_016866603.1
|
2, 3 |
236 |
|
XM_017011115.1 |
XP_016866604.1
|
2, 3 |
336 |
|
XM_017011116.1 |
XP_016866605.1
|
2, 3 |
398 |
|
XM_017011119.1 |
XP_016866608.1
|
2, 3 |
204 |
|
XM_017011120.1 |
XP_016866609.1
|
2, 3 |
395 |
|
XM_017011121.1 |
XP_016866610.1
|
2, 3 |
418 |
|
XM_017011122.1 |
XP_016866611.1
|
2, 3 |
139 |
|
GenBank mRNA |
AK131207.1 |
- |
2, 3 |
418 |
|
AK291760.1 |
- |
2, 3 |
343 |
|
BC034982.2 |
- |
2, 3 |
382 |
4838835 |
DA651352.1 |
- |
2, 3 |
418 |
|
DB094058.1 |
- |
2, 3 |
233 |
|
View Full Product Details