Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020936.2 | NP_065987.1 | 2 | 214 | |
NR_135104.1 | 2 | 214 | |||
NR_135105.1 | 2 | 214 | |||
XM_011538608.2 | XP_011536910.1 | 2 | 202 | ||
XM_017019710.1 | XP_016875199.1 | 2 | 199 | ||
XM_017019711.1 | XP_016875200.1 | 2 | 163 | ||
XM_017019712.1 | XP_016875201.1 | 2 | 52 | ||
XM_017019713.1 | XP_016875202.1 | 2 | 229 | ||
XM_017019714.1 | XP_016875203.1 | 3 | 387 | ||
XM_017019715.1 | XP_016875204.1 | 2 | 200 | ||
XM_017019716.1 | XP_016875205.1 | 2 | 199 | ||
XM_017019717.1 | XP_016875206.1 | 2 | 161 | ||
XM_017019718.1 | XP_016875207.1 | 1 | 671 | ||
XM_017019719.1 | XP_016875208.1 | 2 | 170 | ||
XM_017019720.1 | XP_016875209.1 | 2 | 155 | ||
XM_017019721.1 | XP_016875210.1 | 2 | 200 | ||
GenBank mRNA | AK074219.1 | - | 2 | 166 | |
AK131442.1 | - | 1 | 608 | ||
AK301043.1 | - | 2 | 177 | ||
AK307384.1 | - | 2 | 190 | ||
AL833756.1 | - | 2 | 159 | ||
BC030020.2 | - | 2 | 158 | 4821394 | |
DA806426.1 | - | 2 | 215 |