Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017403.1 | NP_001017403.1 | 18 | 2215 | |
NM_001017404.1 | NP_001017404.1 | 16 | 1798 | ||
NM_021636.2 | NP_067649.2 | 18 | 2102 | ||
XM_005245404.3 | XP_005245461.1 | 18 | 2159 | ||
XM_011509838.2 | XP_011508140.1 | 17 | 2088 | ||
XM_011509839.2 | XP_011508141.1 | 17 | 2089 | ||
XM_011509840.2 | XP_011508142.1 | 17 | 1944 | ||
XM_011509841.2 | XP_011508143.1 | 15 | 1839 | ||
XM_011509842.2 | XP_011508144.1 | 15 | 3567 | ||
XM_011509843.2 | XP_011508145.1 | 15 | 1959 | ||
XM_011509844.2 | XP_011508146.1 | 12 | 1468 | ||
XM_011509846.2 | XP_011508148.1 | 15 | 1864 | ||
XM_017001996.1 | XP_016857485.1 | 16 | 2017 | ||
XM_017001997.1 | XP_016857486.1 | 16 | 1974 | ||
XM_017001998.1 | XP_016857487.1 | 7 | 1314 | ||
GenBank mRNA | AB049405.1 | - | 18 | 2100 | |
AF190501.1 | - | 16 | 1798 | ||
AK027377.1 | - | 1 | 510 | ||
AY358119.1 | - | 18 | 2143 | ||
BC047905.1 | - | 18 | 2085 | 5212508 |