Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282719.1 | NP_001269648.1 | 5 | 718 | |
NM_001282720.1 | NP_001269649.1 | 5 | 700 | ||
NM_001282721.1 | NP_001269650.1 | 5 | 688 | ||
NM_001282722.1 | NP_001269651.1 | 4 | 544 | ||
NM_001282723.1 | NP_001269652.1 | 5 | 609 | ||
NM_001282724.1 | NP_001269653.1 | 4 | 483 | ||
NM_021925.3 | NP_068744.1 | 6 | 844 | ||
NR_104233.1 | 4 | 453 | |||
XM_011533014.2 | XP_011531316.1 | 6 | 871 | ||
GenBank mRNA | AK021672.1 | - | 2 | 66 | |
AK025473.1 | - | 6 | 782 | ||
AK299244.1 | - | 5 | 550 | ||
AK299555.1 | - | 5 | 689 | ||
AK300494.1 | - | 4 | 405 | ||
AK301493.1 | - | 4 | 454 | ||
AK316162.1 | - | 5 | 647 | ||
AK316204.1 | - | 4 | 515 | ||
AK316310.1 | - | 5 | 671 | ||
BC017473.1 | - | 6 | 801 | 3611020 |