Gene Transcripts

Gene Symbol
XPNPEP3
Entrez Gene ID
63929
Gene Name
X-prolyl aminopeptidase 3
Gene Aliases
APP3, ICP55, NPHPL1
Chromosome Location
Chr. 22: 40857081 - 40932819 on Build GRCh38
UniGene ID
Hs.529163
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022098.3 NP_071381.1 3 662
GenBank mRNA AK301635.1 - 3 661
AK310502.1 - 3 920
AK313770.1 - 3 623
AL365514.1 - 3 645
AL834310.1 - 4 704
BC001208.1 - 3 597 3502251
BC001681.2 - 3 629 2823656
BC004989.1 - 3 629 2823656
BX648018.1 - 3 657
CR456442.1 - 3 598
CR457396.1 - 3 561

View Full Product Details