Gene Transcripts

Gene Symbol
GOLPH3
Entrez Gene ID
64083
Gene Name
golgi phosphoprotein 3
Gene Aliases
GOPP1, GPP34, MIDAS, Vps74
Chromosome Location
Chr. 5: 32124711 - 32174319 on Build GRCh38
UniGene ID
Hs.408909
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022130.3 NP_071413.1 1, 2 511
GenBank mRNA AJ296152.1 - 1, 2 332
AK057544.1 - 3, 4 460
AK075156.1 - 1, 2 511
AK092604.1 - 2, 3 372
AK293368.1 - 2, 3 300
AK301588.1 - 2, 3 418
BC012123.1 - 1, 2 466 4558305
BC033725.1 - 1, 2 503 4999629
BC063586.1 - 1, 2 435 4428707

View Full Product Details