Gene Transcripts

Gene Symbol
GREM2
Entrez Gene ID
64388
Gene Name
gremlin 2, DAN family BMP antagonist
Gene Aliases
CKTSF1B2, DAND3, PRDC
Chromosome Location
Chr. 1: 240489573 - 240612372 on Build GRCh38
UniGene ID
Hs.98206
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022469.3 NP_071914.3 2 293
XM_005273226.3 XP_005273283.1 2 121
XM_011544249.2 XP_011542551.1 3 623
GenBank mRNA AK024848.1 - 2 280
BC046632.1 - 2 221 5288555

View Full Product Details