Gene Transcripts

Gene Symbol
DCLRE1B
Entrez Gene ID
64858
Gene Name
DNA cross-link repair 1B
Gene Aliases
APOLLO, SNM1B, SNMIB
Chromosome Location
Chr. 1: 113904619 - 113914086 on Build GRCh38
UniGene ID
Hs.591412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022836.3 NP_073747.1 2 520
GenBank mRNA AF306693.1 - 2 455
AK022872.1 - 2 66
AK024060.1 - 2 438
BC029687.1 - 2 397 5001181

View Full Product Details