Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031903.2 | NP_114109.1 | 2 | 215 | |
GenBank mRNA | AB049649.1 | - | 2 | 214 | |
AF161401.1 | - | 2 | 155 | ||
AK090551.1 | - | 2 | 179 | ||
AK313393.1 | - | 2 | 173 | ||
BC013147.1 | - | 2 | 174 | 4340082 | |
BM833110.1 | - | 2 | 178 | ||
DB456673.1 | - | 2 | 357 |