Gene Transcripts

Gene Symbol
GCC1
Entrez Gene ID
79571
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
GCC1P, GCC88
Chromosome Location
Chr. 7: 127580628 - 127585600 on Build GRCh38
UniGene ID
Hs.521168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024523.5 NP_078799.2 2 1896
GenBank mRNA AF525417.1 - 2 1671
AK025688.1 - 2 555
AK092878.1 - 2 1352
AK315472.1 - 2 1747
AL832833.1 - 2 558
BC008902.2 - 2 678
BC014100.2 - 2 1896 2988591
BC078665.1 - 2 1848 5923117

View Full Product Details