Gene Transcripts

Gene Symbol
MSANTD2
Entrez Gene ID
79684
Gene Name
Myb/SANT DNA binding domain containing 2
Gene Aliases
C11orf61
Chromosome Location
Chr. 11: 124766492 - 124800879 on Build GRCh38
UniGene ID
Hs.146079
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308027.1 NP_001294956.1 2 713
NM_001312920.1 NP_001299849.1 1 1447
NM_001312921.1 NP_001299850.1 2 570
NM_024631.3 NP_078907.2 2 557
XM_005271674.3 XP_005271731.3 3 1122
XM_006718905.1 XP_006718968.1 2 2082
XM_006718906.3 XP_006718969.1 2 1918
XM_011542981.2 XP_011541283.2 2 1188
XM_011542982.2 XP_011541284.2 2 1188
XM_017018295.1 XP_016873784.1 2 17328
XM_017018296.1 XP_016873785.1 2 2238
XM_017018297.1 XP_016873786.1 3 539
XM_017018298.1 XP_016873787.1 3 13299
XM_017018299.1 XP_016873788.1 3 340
GenBank mRNA AK026995.1 - 2 556
AK092413.1 - 1 1121
AL512691.1 - 1 1447
BC064919.1 - 2 698 6093009
CX781748.1 - 2 556
DA379833.1 - 2 557

View Full Product Details