Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193478.1 | NP_001180407.1 | 4 | 461 | |
NM_030911.3 | NP_112173.1 | 4 | 461 | ||
XM_011535250.2 | XP_011533552.1 | 4 | 425 | ||
XM_011535251.1 | XP_011533553.1 | 4 | 405 | ||
XM_017020779.1 | XP_016876268.1 | 4 | 453 | ||
XM_017020780.1 | XP_016876269.1 | 4 | 803 | ||
XM_017020782.1 | XP_016876271.1 | 3 | 251 | ||
GenBank mRNA | AK312835.1 | - | 4 | 431 | |
AY027525.1 | - | 4 | 410 | ||
BC032889.1 | - | 4 | 436 | ||
BC070238.1 | - | 4 | 429 | ||
BC092434.1 | - | 4 | 439 | ||
BC125202.1 | - | 4 | 375 | 40123430 | |
DA400049.1 | - | 4 | 461 | ||
DA560379.1 | - | 4 | 461 |