Gene Transcripts
- Gene Symbol
- ACSBG2
- Entrez Gene ID
- 81616
- Gene Name
- acyl-CoA synthetase bubblegum family member 2
- Gene Aliases
- BGR, BRGL, PRTD-NY3, PRTDNY3
- Chromosome Location
- Chr. 19: 6135633 - 6193103 on Build GRCh38
- UniGene ID
- Hs.465720
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289177.1 |
NP_001276106.1
|
11, 12 |
1853 |
|
NM_001289178.1 |
NP_001276107.1
|
11, 12 |
1799 |
|
NM_001289179.1 |
NP_001276108.1
|
11, 12 |
1844 |
|
NM_001289180.1 |
NP_001276109.1
|
11, 12 |
1563 |
|
NM_001321384.1 |
NP_001308313.1
|
11, 12 |
1834 |
|
NM_030924.4 |
NP_112186.3
|
11, 12 |
1844 |
|
XM_005259653.3 |
XP_005259710.1
|
11, 12 |
1826 |
|
XM_011528329.1 |
XP_011526631.1
|
8, 9 |
1270 |
|
XM_017027332.1 |
XP_016882821.1
|
11, 12 |
1671 |
|
XM_017027333.1 |
XP_016882822.1
|
11, 12 |
1826 |
|
XM_017027334.1 |
XP_016882823.1
|
10, 11 |
1658 |
|
GenBank mRNA |
AJ577571.1 |
- |
11, 12 |
1747 |
|
AK057412.1 |
- |
6, 7 |
1055 |
|
AK093433.1 |
- |
11, 12 |
1812 |
|
AK225862.1 |
- |
10, 11 |
1552 |
|
AK302604.1 |
- |
11, 12 |
1801 |
|
AL136854.1 |
- |
11, 12 |
1585 |
|
AY009107.1 |
- |
11, 12 |
1556 |
|
AY358766.1 |
- |
11, 12 |
1844 |
|
BC022027.1 |
- |
11, 12 |
1775 |
4825681 |
BC090046.1 |
- |
11, 12 |
1760 |
30388900 |
View Full Product Details