Gene Transcripts

Gene Symbol
SF3B5
Entrez Gene ID
83443
Gene Name
splicing factor 3b subunit 5
Gene Aliases
SF3b10, Ysf3
Chromosome Location
Chr. 6: 144094881 - 144095617 on Build GRCh38
UniGene ID
Hs.110695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031287.2 NP_112577.1 1 316
GenBank mRNA AK074417.1 - 1 272
AK312079.1 - 1 271
BC000198.1 - 1 256 3352960

View Full Product Details