Gene Transcripts
- Gene Symbol
- CDCA7
- Entrez Gene ID
- 83879
- Gene Name
- cell division cycle associated 7
- Gene Aliases
- ICF3, JPO1
- Chromosome Location
- Chr. 2: 173354017 - 173368992 on Build GRCh38
- UniGene ID
- Hs.470654
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031942.4 |
NP_114148.3
|
7, 8 |
1171 |
|
NM_145810.2 |
NP_665809.1
|
6, 7 |
934 |
|
XM_011511957.2 |
XP_011510259.1
|
7, 8 |
1712 |
|
GenBank mRNA |
AK027628.1 |
- |
7, 8 |
1143 |
|
AK027642.1 |
- |
6, 7 |
915 |
|
AK075134.1 |
- |
5, 6 |
1797 |
|
AK223524.1 |
- |
6, 7 |
939 |
|
AK297097.1 |
- |
5, 6 |
769 |
|
AK297245.1 |
- |
7, 8 |
1031 |
|
AK300949.1 |
- |
7, 8 |
1000 |
|
AL833728.1 |
- |
2, 3 |
463 |
|
AY029179.1 |
- |
6, 7 |
947 |
|
BC015124.1 |
- |
2, 3 |
350 |
|
BC027966.1 |
- |
6, 7 |
900 |
5227112 |
View Full Product Details