Gene Transcripts

Gene Symbol
TSSK6
Entrez Gene ID
83983
Gene Name
testis specific serine kinase 6
Gene Aliases
CT72, FKSG82, SSTK, TSSK4
Chromosome Location
Chr. 19: 19514219 - 19515660 on Build GRCh38
UniGene ID
Hs.532711
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032037.3 NP_114426.1 1 292
GenBank mRNA AF329483.1 - 1 181
AF348077.1 - 1 317
AK058166.1 - 1 180
AK172841.1 - 1 661
AK313767.1 - 1 180

View Full Product Details