Gene Transcripts

Gene Symbol
SPNS1
Entrez Gene ID
83985
Gene Name
sphingolipid transporter 1 (putative)
Gene Aliases
HSpin1, LAT, PP2030, SPIN1, SPINL, nrs
Chromosome Location
Chr. 16: 28974767 - 28984769 on Build GRCh38
UniGene ID
Hs.617449
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142448.1 NP_001135920.1 3 400
NM_001142450.1 NP_001135922.1 3 550
NM_001142451.1 NP_001135923.1 2 637
NM_032038.2 NP_114427.1 2 637
XM_006721096.3 XP_006721159.1 3 407
XM_017023757.1 XP_016879246.1 3 408
XM_017023758.1 XP_016879247.1 3 404
XM_017023759.1 XP_016879248.1 3 403
GenBank mRNA AF212371.1 - 2 584
AK095677.1 - 2 634
AK289787.1 - 2 632
AL524908.3 - 3 525
BC006156.1 - 2 248
BC008325.1 - 2 462 3501826
BC038961.1 - 3 414 6053095
BC047741.1 - 2 611 5757339
BC065235.1 - 3 382 6161778
BG771918.1 - 2 645

View Full Product Details