Gene Transcripts

Gene Symbol
LRCH3
Entrez Gene ID
84859
Gene Name
leucine rich repeats and calponin homology domain containing 3
Gene Aliases
Chromosome Location
Chr. 3: 197790855 - 197889346 on Build GRCh38
UniGene ID
Hs.518414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032773.3 NP_116162.1 6 876
XM_005269362.2 XP_005269419.1 6 1247
XM_005269363.2 XP_005269420.1 6 1247
XM_005269365.2 XP_005269422.1 6 1247
XM_005269367.2 XP_005269424.1 6 1247
XM_006713791.3 XP_006713854.1 6 1247
XM_006713792.3 XP_006713855.1 6 1247
XM_006713793.2 XP_006713856.1 6 1247
XM_011513242.2 XP_011511544.1 6 1247
XM_011513243.2 XP_011511545.1 6 1247
XM_017007350.1 XP_016862839.1 6 1247
XM_017007351.1 XP_016862840.1 6 1247
XM_017007352.1 XP_016862841.1 6 1247
GenBank mRNA AK293745.1 - 6 865
AK300466.1 - 3 460
AK303521.1 - 6 838
BC007504.2 - 6 828 2960711

View Full Product Details