Gene Transcripts

Gene Symbol
SPSB3
Entrez Gene ID
90864
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
C16orf31, SSB3
Chromosome Location
Chr. 16: 1776712 - 1782605 on Build GRCh38
UniGene ID
Hs.592080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324081.1 NP_001311010.1 6, 7 780
NM_080861.3 NP_543137.2 6, 7 802
XM_005255673.4 XP_005255730.1 6, 7 992
GenBank mRNA AF403028.1 - 5, 6 711
AK057850.1 - 560
AK074190.1 - 6, 7 766
BC007588.1 - 6, 7 748 3342825
BC024244.1 - 6, 7 766 4798873
BC035065.1 - 6, 7 759
BC041897.1 - 5, 6 1576 5298729
BC047441.1 - 6, 7 744 5303706
BC065515.1 - 3, 4 1045
BI768095.1 - 6, 7 756
BQ930544.1 - 1, 2 84

View Full Product Details