Gene Transcripts

Gene Symbol
ANKRD30A
Entrez Gene ID
91074
Gene Name
ankyrin repeat domain 30A
Gene Aliases
NY-BR-1
Chromosome Location
Chr. 10: 37125754 - 37235831 on Build GRCh38
UniGene ID
Hs.373787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052997.2 NP_443723.2 21 2162
XM_011519757.2 XP_011518059.1 22 2307
XM_011519758.2 XP_011518060.1 22 2307
XM_017016883.1 XP_016872372.1 3 178
GenBank mRNA AF269087.1 - 21 2162
BP312910.1 - 3 193

View Full Product Details