Gene Transcripts

Gene Symbol
ELMSAN1
Entrez Gene ID
91748
Gene Name
ELM2 and Myb/SANT domain containing 1
Gene Aliases
C14orf117, C14orf43, LSR68, MIDEAS, c14_5541
Chromosome Location
Chr. 14: 73715122 - 73790285 on Build GRCh38
UniGene ID
Hs.594157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001043318.2 NP_001036783.1 2 1526
NM_194278.3 NP_919254.2 2 1905
XM_005268204.1 XP_005268261.1 2 1905
XM_005268205.1 XP_005268262.1 2 1461
XM_005268206.1 XP_005268263.1 3 1678
GenBank mRNA AK127785.1 - 2 1904
BC003121.1 - 1 588
BC006451.1 - 1 360
BC006511.2 - 1 1156
BC009202.2 - 2 1526
BC015668.2 - 2 1539
BC025330.1 - 1 1156

View Full Product Details