Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080660.3 | NP_542391.2 | 3 | 552 | |
XM_006716176.3 | XP_006716239.1 | 3 | 995 | ||
XM_011516688.2 | XP_011514990.1 | 3 | 993 | ||
XM_011516689.2 | XP_011514991.1 | 3 | 897 | ||
GenBank mRNA | BC008842.2 | - | 3 | 552 |