Gene Transcripts

Gene Symbol
DPH7
Entrez Gene ID
92715
Gene Name
diphthamide biosynthesis 7
Gene Aliases
C9orf112, RRT2, WDR85
Chromosome Location
Chr. 9: 137554899 - 137578935 on Build GRCh38
UniGene ID
Hs.292570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138778.3 NP_620133.1 9 1295
XM_005266123.2 XP_005266180.1 10 1753
XM_005266126.3 XP_005266183.1 8 1526
XM_005266128.2 XP_005266185.1 10 1379
XM_005266129.2 XP_005266186.1 10 1382
XM_005266130.2 XP_005266187.1 8 1150
XM_011519188.1 XP_011517490.1 8 1218
XM_011519189.1 XP_011517491.1 9 1396
XM_011519191.2 XP_011517493.1 9 2451
XM_011519193.2 XP_011517495.1 9 2500
XM_011519199.2 XP_011517501.1 9 1241
XM_011519200.1 XP_011517502.1 9 1244
XM_017015289.1 XP_016870778.1 8 2385
XM_017015290.1 XP_016870779.1 8 2434
XM_017015291.1 XP_016870780.1 10 1588
XM_017015292.1 XP_016870781.1 9 1658
XM_017015293.1 XP_016870782.1 9 1615
XM_017015294.1 XP_016870783.1 11 1568
XM_017015295.1 XP_016870784.1 9 1655
XM_017015296.1 XP_016870785.1 9 1494
XM_017015297.1 XP_016870786.1 11 1565
XM_017015298.1 XP_016870787.1 10 1848
XM_017015299.1 XP_016870788.1 10 1105
XM_017015300.1 XP_016870789.1 10 1845
XM_017015301.1 XP_016870790.1 8 1518
XM_017015302.1 XP_016870791.1 8 2572
XM_017015303.1 XP_016870792.1 9 1753
XM_017015304.1 XP_016870793.1 8 1218
XM_017015305.1 XP_016870794.1 8 2549
XM_017015306.1 XP_016870795.1 9 1687
XM_017015307.1 XP_016870796.1 9 1684
XM_017015308.1 XP_016870797.1 7 2411
XM_017015309.1 XP_016870798.1 9 1411
GenBank mRNA AJ420445.1 - 1 654
AK075115.1 - 9 1296
AK094389.1 - 1 1296
BC003123.1 - 9 1234 3502107
BC017335.2 - 9 1254 4642600
BC040173.1 - 2 757 4791980

View Full Product Details