Gene Transcripts
- Gene Symbol
- LEMD1
- Entrez Gene ID
- 93273
- Gene Name
- LEM domain containing 1
- Gene Aliases
- CT50, LEMP-1
- Chromosome Location
- Chr. 1: 205381378 - 205449956 on Build GRCh38
- UniGene ID
- Hs.655520
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001001552.4 |
NP_001001552.3
|
2, 3 |
227 |
|
NM_001199050.1 |
NP_001185979.1
|
2, 3 |
227 |
|
XM_011510160.2 |
XP_011508462.1
|
2, 3 |
231 |
|
XM_011510161.2 |
XP_011508463.1
|
2, 3 |
219 |
|
XM_011510162.2 |
XP_011508464.1
|
2, 3 |
229 |
|
XM_011510164.2 |
XP_011508466.1
|
2, 3 |
251 |
|
XM_011510165.2 |
XP_011508467.1
|
2, 3 |
247 |
|
GenBank mRNA |
AB085610.1 |
- |
2, 3 |
194 |
|
AB096677.1 |
- |
2, 3 |
194 |
|
AB096680.1 |
- |
2, 3 |
194 |
|
AB096681.1 |
- |
2, 3 |
194 |
|
AK026628.1 |
- |
2, 3 |
194 |
|
AK302392.1 |
- |
2, 3 |
229 |
|
BC036636.2 |
- |
2, 3 |
197 |
5296437 |
DC399934.1 |
- |
2, 3 |
229 |
|
View Full Product Details